Novus Biologicals
Manufacturer Code:NBP203794
Catalog # NBP203794
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D6 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 284-669 of human TRIM9 (NP_443210) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM9; EC 6.3.2.- KIAA0282homolog of rat RING finger Spring RNF91RING finger protein 91 SPRINGE3 ubiquitin-protein ligase TRIM9 tripartite motif containing 9 tripartite motif-containing 9 Tripartite motif-containing protein 9; homolog of rat RING finger Spring; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; tripartite motif-containing 9; Tripartite motif-containing protein 9
Gene Aliases: KIAA0282; RNF91; SPRING; TRIM9
UniProt ID: (Human) Q9C026
Entrez Gene ID: (Human) 114088
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.