Invitrogen
Manufacturer Code:PA540853
Catalog # PIPA540853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human TRIM9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM9; homolog of rat RING finger Spring; RING finger protein 91; RING-type E3 ubiquitin transferase TRIM9; SNAP-25-interacting RING finger protein; Spring; Tripartite motif containing 9 KIAA0282 RNF91 SPRING; tripartite motif protein 9; tripartite motif-containing 9; Tripartite motif-containing protein 9
Gene Aliases: fj48c11; KIAA0282; RNF91; SPRING; TRIM9; wu:fj48c11; zgc:77357
UniProt ID: (Human) Q9C026, (Bovine) Q29RQ5, (Mouse) Q8C7M3, (Rat) Q91ZY8
Entrez Gene ID: (Human) 114088, (Rabbit) 100351846, (Dog) 490687, (Horse) 100066749, (Bovine) 767615, (Mouse) 94090, (Guinea pig) 100729922, (Rat) 155812, (Zebrafish) 336099
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.