Proteintech
Manufacturer Code:221511AP150UL
Catalog # 50-173-3746
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIM72 Fusion Protein Ag17756 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hMG53 Mg53 Mitsugumin 53 TRIM72 tripartite motif containing 72; Mg53; mitsugumin 53; Mitsugumin-53; TRIM72; tripartite motif containing 72, E3 ubiquitin protein ligase; tripartite motif-containing 72; Tripartite motif-containing protein 72
Gene Aliases: BC067209; MG53; TRIM72
UniProt ID: (Human) Q6ZMU5, (Mouse) Q1XH17
Entrez Gene ID: (Human) 493829, (Mouse) 434246
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.