Proteintech
Manufacturer Code:554561AP150UL
Catalog # 50-173-6170
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM63; IRF Iris RING finger protein MuRF 1 MURF1 MURF2 RING finger protein 28 RNF28 SMRZ TRIM63 tripartite motif containing 63; Iris RING finger protein; MURF-1; muscle ring finger protein 1; muscle specific ring finger protein 2; Muscle-specific RING finger protein 1; RING finger protein 28; RING-type E3 ubiquitin transferase TRIM63; Striated muscle RING zinc finger protein; tripartite motif containing 63, E3 ubiquitin protein ligase; tripartite motif protein 63; tripartite motif-containing 63; Tripartite motif-containing protein 63
Gene Aliases: IRF; Murf; MURF1; MURF2; RF1; RNF28; SMRZ; TRIM63
UniProt ID: (Human) Q969Q1, (Rat) Q91Z63
Entrez Gene ID: (Human) 84676, (Rat) 140939, (Mouse) 433766
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.