Invitrogen
Manufacturer Code:MA527076
Catalog # PIMA527076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8E12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 311-554 of TRIM56 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM56; RING finger protein 109; RING-type E3 ubiquitin transferase TRIM56; RNF109 E3 ubiquitin-protein ligase TRIM56 RING finger protein 109 tripartite motif-containing 56 tripartite motif-containing protein 56; Tripartite motif-containing protein 56
Gene Aliases: RNF109; TRIM56
UniProt ID: (Human) Q9BRZ2
Entrez Gene ID: (Human) 81844
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.