Novus Biologicals
Manufacturer Code:NBP180372
Catalog # NBP180372
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TRIM56. Peptide sequence DPKGSLLGDFLTAYHGLEKPRVTTMVDGRYLVVSLSNGTIHIFRVRSPDS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp667O116 E3 ubiquitin-protein ligase TRIM56 EC 6.3.2.- RING finger protein 109 RNF109FLJ35608 tripartite motif containing 56 tripartite motif-containing 56 Tripartite motif-containing protein 56; E3 ubiquitin-protein ligase TRIM56; RING finger protein 109; RING-type E3 ubiquitin transferase TRIM56; Tripartite motif-containing protein 56
Gene Aliases: RNF109; TRIM56
UniProt ID: (Human) Q9BRZ2
Entrez Gene ID: (Human) 81844
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.