Invitrogen
Manufacturer Code:MA519380
Catalog # PIMA519380
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D11 |
Immunogen | TRIM37 (NP&lowbar056109 865 a.a. &bsim 964 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM37; MUL POB1 TEF3 E3 ubiquitin-protein ligase TRIM37 RING-B-box-coiled-coil protein mulibrey ism protein tripartite motif-containing 37 tripartite motif-containing protein 37; Mulibrey nanism protein; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; Tripartite motif-containing protein 37
Gene Aliases: KIAA0898; MUL; POB1; TEF3; TRIM37
UniProt ID: (Human) O94972
Entrez Gene ID: (Human) 4591
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.