Novus Biologicals
Manufacturer Code:NB10059821
Catalog # NB10059821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 914 and 964 of human tripartite motif-containing 37 using the numbering given in entry NP_056109.1 (GeneID 4591). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM37; EC 6.3.2.- KIAA0898Tripartite motif-containing protein 37 Mulibrey nanism protein MULRING-B-box-coiled-coil protein POB1E3 ubiquitin-protein ligase TRIM37 TEF3 tripartite motif containing 37 tripartite motif-containing 37; Mulibrey nanism protein; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; Tripartite motif-containing protein 37
Gene Aliases: KIAA0898; MUL; POB1; TEF3; TRIM37
UniProt ID: (Human) O94972
Entrez Gene ID: (Human) 4591
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.