Proteintech
Manufacturer Code:130371AP150UL
Catalog # 50-172-8437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIM37 Fusion Protein Ag3690 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase TRIM37; KIAA0898 MUL Mulibrey ism protein POB1 TEF3 TRIM37 tripartite motif containing 37; Mulibrey nanism protein; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TRAF encompassing factor 3; tripartite motif protein 37; Tripartite motif-containing protein 37
Gene Aliases: 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; KIAA0898; mKIAA0898; MUL; POB1; TEF3; TRIM37
UniProt ID: (Human) O94972, (Mouse) Q6PCX9
Entrez Gene ID: (Human) 4591, (Mouse) 68729, (Rat) 360592
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.