Invitrogen
Manufacturer Code:PA5110518
Catalog # PIPA5110518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TRIM32 (NP_036342.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810045E12Rik 3f3 72 kDa Tat-interacting protein BBS11 BOS_9387 E3 ubiquitin-protein ligase TRIM32 HT2A LGMD2H RING-type E3 ubiquitin transferase TRIM32 RP11-67K19.1 TATIP Trim32 Tripartite motif-containing protein 32 Zfp117 Zinc finger protein HT2A; 72 kDa Tat-interacting protein; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; RING-type E3 ubiquitin transferase TRIM32; TAT-interactive protein, 72-KD; tripartite motif protein 32; tripartite motif-containing 32; Tripartite motif-containing protein 32; zinc finger protein 117; Zinc finger protein HT2A; zinc-finger protein HT2A
Gene Aliases: 1810045E12Rik; 3f3; BBS11; HT2A; LGMD2H; TATIP; TRIM32; Zfp117
UniProt ID: (Human) Q13049, (Mouse) Q8CH72
Entrez Gene ID: (Human) 22954, (Mouse) 69807, (Rat) 313264
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.