R&D Systems
Manufacturer Code:AF6515SP
Catalog # AF6515SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human TRIM32 Arg105-Lys204 Accession # Q13049 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 72 kDa Tat-interacting protein; 72 kDa Tat-interacting protein BBS11 BBS1172-KD EC 6.3.2.- HT2A HT2Alimb girdle muscular dystrophy 2H (autosomal recessive) LGMD2H TATIP tripartite motif containing 32 Zinc finger protein HT2A zinc-finger protein HT2A; E3 ubiquitin-protein ligase TRIM32; RING-type E3 ubiquitin transferase TRIM32; TAT-interactive protein, 72-KD; tripartite motif-containing 32; Tripartite motif-containing protein 32; Zinc finger protein HT2A; zinc-finger protein HT2A
Gene Aliases: BBS11; HT2A; LGMD2H; TATIP; TRIM32
UniProt ID: (Human) Q13049
Entrez Gene ID: (Human) 22954
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.