Novus Biologicals
Manufacturer Code:NBP248093
Catalog # NBP248093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TRIM29/1041 |
Immunogen | Recombinant fragment (126 Amino acid residues between aa 1-200) of human TRIM29 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ataxia telangiectasia group D-associated protein; Ataxia telangiectasia group D-associated protein ATDCataxia-telangiectasia group D-associated protein FLJ36085 tripartite motif containing 29 tripartite motif protein TRIM29 tripartite motif-containing 29 tripartite motif-containing protein 29; ataxia-telangiectasia group D-associated protein; tripartite motif protein TRIM29; tripartite motif-containing 29; Tripartite motif-containing protein 29
Gene Aliases: ATDC; TRIM29
UniProt ID: (Human) Q14134
Entrez Gene ID: (Human) 23650
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.