Invitrogen
Manufacturer Code:PA577239
Catalog # PIPA577239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human TRIM29. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ataxia telangiectasia group D-associated protein; ataxia-telangiectasia group D-associated protein; tripartite motif protein 29; tripartite motif protein TRIM29; tripartite motif-containing 29; Tripartite motif-containing protein 29; Tripartite motif-containing protein 29 Ataxia telangiectasia group D-associated protein TRIM29 ATDC
Gene Aliases: 1110047J21Rik; 2810431N19Rik; 4732461M22Rik; AI119726; ATDC; TRIM29
UniProt ID: (Human) Q14134, (Mouse) Q8R2Q0
Entrez Gene ID: (Human) 23650, (Mouse) 72169, (Rat) 300656
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.