Novus Biologicals
Manufacturer Code:NB12022207
Catalog # NB12022207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against synthetic peptides corresponding to amino acids 569-583 of human TRIM29. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ataxia telangiectasia group D-associated protein; Ataxia telangiectasia group D-associated protein ATDCataxia-telangiectasia group D-associated protein FLJ36085 tripartite motif containing 29 tripartite motif protein TRIM29 tripartite motif-containing 29 tripartite motif-containing protein 29; ataxia-telangiectasia group D-associated protein; tripartite motif protein TRIM29; tripartite motif-containing 29; Tripartite motif-containing protein 29
Gene Aliases: ATDC; TRIM29
UniProt ID: (Human) Q14134
Entrez Gene ID: (Human) 23650
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.