Invitrogen
Manufacturer Code:MA535303
Catalog # PIMA535303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0047 |
Immunogen | A synthesized peptide derived from human KAP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AA408787 E3 SUMO-protein ligase TRIM28 KAP1 KAP-1 KRAB-A-interacting protein KRAB-associated protein 1 KRAB-interacting p KRAB-interacting protein 1 Krip1 KRIP-1 MommeD9 Nuclear corepressor KAP-1 PPP1R157 RING finger protein 96 RING-type E3 ubiquitin transferase TIF1-beta RNF96 TF1B Tif1b Tif1beta TIF1-beta Transcription intermediary factor 1-beta TRIM28 tripartite motif Tripartite motif-containing protein 28; E3 SUMO-protein ligase TRIM28; KAP-1; KRAB [Kruppel-associated box domain]-associated protein 1; KRAB-A-interacting protein; KRAB-associated protein 1; KRAB-interacting protein 1; KRIP-1; Nuclear corepressor KAP-1; protein phosphatase 1, regulatory subunit 157; RING finger protein 96; RING-type E3 ubiquitin transferase TIF1-beta; TIF1-beta; Transcription intermediary factor 1-beta; transcriptional intermediary factor 1, beta; transcriptional intermediary factor 1-beta; tripartite motif protein 28; Tripartite motif-containing protein 28
Gene Aliases: AA408787; KAP-1; KAP1; KRIP-1; Krip1; MommeD9; PPP1R157; RNF96; TF1B; TIF1B; Tif1beta; TRIM28
UniProt ID: (Human) Q13263, (Mouse) Q62318, (Rat) O08629
Entrez Gene ID: (Human) 10155, (Mouse) 21849, (Rat) 116698
Molecular Function:
transcription cofactor
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.