Novus Biologicals
Manufacturer Code:NBP246214
Catalog # NBP246214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D4 |
Immunogen | Full length human recombinant protein of human TRIM27 (NP_006501) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ret finger protein; Ret finger proteinRING finger protein 76 RFPtripartite motif protein TRIM27 RNF76RFP transforming protein tripartite motif containing 27 tripartite motif-containing 27 Tripartite motif-containing protein 27 zinc finger protein RFP; RFP transforming protein; RING finger protein 76; RING-type E3 ubiquitin transferase TRIM27; tripartite motif protein TRIM27; tripartite motif-containing 27; Tripartite motif-containing protein 27; Zinc finger protein RFP
Gene Aliases: RFP; RNF76; TRIM27
UniProt ID: (Human) P14373
Entrez Gene ID: (Human) 5987
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.