Proteintech
Manufacturer Code:270131AP150UL
Catalog # 50-173-5532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIM26 Fusion Protein Ag25546 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid finger protein; Acid finger protein AFP RING finger protein 95 RNF95 TRIM26 tripartite motif containing 26 Zinc finger protein 173 ZNF173; AFP; RING finger protein 95; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; Tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zinc finger protein 173
Gene Aliases: AFP; AI462198; RNF95; TRIM26; Zfp173; Zfp1736; ZNF173
UniProt ID: (Human) Q12899, (Mouse) Q99PN3, (Rat) P62603
Entrez Gene ID: (Human) 7726, (Mouse) 22670, (Rat) 309586
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.