Novus Biologicals
Manufacturer Code:NBP261814
Catalog # NBP2-61814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5B5B12 |
Immunogen | Purified recombinant fragment of human TRIM25 (AA: 211-360) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: E3 ubiquitin/ISG15 ligase TRIM25; EC 6.3.2.19 EC 6.3.2.n3 EFPZ147 Estrogen-responsive finger protein RING finger protein 147 RNF147E3 ubiquitin/ISG15 ligase TRIM25 tripartite motif containing 25 tripartite motif-containing 25 Tripartite motif-containing protein 25 Ubiquitin/ISG15-conjugating enzyme TRIM25 Zinc finger protein 147 zinc finger protein 147 (estrogen-responsive finger protein) zinc finger protein-147 ZNF147tripartite motif protein TRIM25; Estrogen-responsive finger protein; RING finger protein 147; RING-type E3 ubiquitin transferase; RING-type E3 ubiquitin transferase TRIM25; tripartite motif protein TRIM25; tripartite motif-containing 25; Tripartite motif-containing protein 25; Ubiquitin/ISG15-conjugating enzyme TRIM25; Zinc finger protein 147; zinc finger protein 147 (estrogen-responsive finger protein); zinc finger protein-147
Gene Aliases: EFP; RNF147; TRIM25; Z147; ZNF147
UniProt ID: (Human) Q14258
Entrez Gene ID: (Human) 7706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.