Invitrogen
Manufacturer Code:PA520410
Catalog # PIPA520410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide from near the carboxy terminus of human TRIM25. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin/ISG15 ligase TRIM25; EFP RNF147 Z147 ZNF147 E3 ubiquitin/ISG15 ligase TRIM25 RING finger protein 147 estrogen-responsive finger protein tripartite motif protein TRIM25 tripartite motif-containing 25 tripartite motif-containing protein 25 ubiquitin/ISG15-conjugating enzyme TRIM25 zinc finger protein 147 (estrogen-responsive finger protein) zinc finger protein-147; Estrogen-responsive finger protein; RING finger protein 147; RING-type E3 ubiquitin transferase; RING-type E3 ubiquitin transferase TRIM25; tripartite motif protein 25; tripartite motif protein TRIM25; tripartite motif-containing 25; Tripartite motif-containing protein 25; Ubiquitin/ISG15-conjugating enzyme TRIM25; Zinc finger protein 147; zinc finger protein 147 (estrogen-responsive finger protein); zinc finger protein-147
Gene Aliases: AA960166; AL022677; EFP; RNF147; TRIM25; Z147; Zfp147; ZNF147
UniProt ID: (Human) Q14258, (Mouse) Q61510
Entrez Gene ID: (Human) 7706, (Mouse) 217069
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.