Invitrogen
Manufacturer Code:PA5110701
Catalog # PIPA5110701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human TRIM25 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960166 AL022677 E3 ubiquitin/ISG15 ligase TRIM25 Efp Estrogen-responsive finger protein RING finger protein 147 RING-type E3 ubiquitin transferase RING-type E3 ubiquitin transferase TRIM25 RNF147 Trim25 Tripartite motif-containing protein 25 Ubiquitin/ISG15-conjugating enzyme TRIM25 Z147 Zfp147 Zinc finger protein 147 Znf147; E3 ubiquitin/ISG15 ligase TRIM25; Estrogen-responsive finger protein; RING finger protein 147; RING-type E3 ubiquitin transferase; RING-type E3 ubiquitin transferase TRIM25; tripartite motif protein TRIM25; tripartite motif-containing 25; Tripartite motif-containing protein 25; Ubiquitin/ISG15-conjugating enzyme TRIM25; Zinc finger protein 147; zinc finger protein 147 (estrogen-responsive finger protein); zinc finger protein-147
Gene Aliases: EFP; RNF147; TRIM25; Z147; ZNF147
UniProt ID: (Human) Q14258
Entrez Gene ID: (Human) 7706
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.