Proteintech
Manufacturer Code:125731AP150UL
Catalog # 50-172-8078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIM25 Fusion Protein Ag3241 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E3 ubiquitin/ISG15 ligase TRIM25; EFP RING finger protein 147 RNF147 TRIM25 tripartite motif containing 25 Z147 Zinc finger protein 147 ZNF147; Estrogen-responsive finger protein; RING finger protein 147; RING-type E3 ubiquitin transferase; RING-type E3 ubiquitin transferase TRIM25; tripartite motif protein 25; tripartite motif protein TRIM25; tripartite motif-containing 25; Tripartite motif-containing protein 25; Ubiquitin/ISG15-conjugating enzyme TRIM25; Zinc finger protein 147; zinc finger protein 147 (estrogen-responsive finger protein); zinc finger protein-147
Gene Aliases: AA960166; AL022677; EFP; RNF147; TRIM25; Z147; Zfp147; ZNF147
UniProt ID: (Human) Q14258, (Mouse) Q61510
Entrez Gene ID: (Human) 7706, (Mouse) 217069, (Rat) 494338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.