Novus Biologicals
Manufacturer Code:NBP30380920UL
Catalog # NB092648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 80-330 of human TRIM21/SSA1 (NP_003132.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa ribonucleoprotein autoantigen Ro/SS-A 52 kDa Ro/SSA E3 ubiquitin-protein ligase TRIM21 EC 6.3.2.- rhTRIM21 RNF81RING finger protein 81 ro(SS-A) RO52SS-A Sicca syndrome antigen A Sjoegren syndrome type A antigen Sjogren syndrome antigen A1 (52kDa ribonucleoprotein autoantigen SS-A/Ro) SSA1SSA tripartite motif containing 2152 kDa Ro protein tripartite motif-containing 21 Tripartite motif-containing protein 21; 52 kDa Ro protein; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; Ro(SS-A); Sicca syndrome antigen A; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SS-A; tripartite motif-containing 21; Tripartite motif-containing protein 21
Gene Aliases: RNF81; Ro/SSA; RO52; SSA; SSA1; TRIM21
UniProt ID: (Human) P19474
Entrez Gene ID: (Human) 6737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.