R&D Systems
Manufacturer Code:AF6219
Catalog # AF6219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human TRIM21 Met1-Tyr475 Accession # P19474 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa ribonucleoprotein autoantigen Ro/SS-A 52 kDa Ro/SSA E3 ubiquitin-protein ligase TRIM21 EC 6.3.2.- rhTRIM21 RNF81 RNF81RING finger protein 81 ro(SS-A) Ro52 RO52SS-A Sicca syndrome antigen A Sjoegren syndrome type A antigen Sjogren syndrome antigen A1 (52kDa ribonucleoprotein autoantigen SS-A/Ro) SSA1 SSA1SSA tripartite motif containing 2152 kDa Ro protein tripartite motif-containing 21 Tripartite motif-containing protein 21; 52 kDa Ro protein; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; Ro(SS-A); Sicca syndrome antigen A; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SS-A; tripartite motif-containing 21; Tripartite motif-containing protein 21
Gene Aliases: RNF81; Ro/SSA; RO52; SSA; SSA1; TRIM21
UniProt ID: (Human) P19474
Entrez Gene ID: (Human) 6737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.