Invitrogen
Manufacturer Code:PA522294
Catalog # PIPA522294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 254 and 475 of SSA1 (Uniprot IDNo.P19474) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa Ro protein; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; RNF81 RO52 SSA SSA1 52 kDa Ro protein 52 kDa ribonucleoprotein autoantigen Ro/SS-A E3 ubiquitin-protein ligase TRIM21 RING finger protein 81 SS-A Sicca syndrome antigen A Sjogren syndrome antigen A1 (52kDa ribonucleoprotein autoantigen SS-A/Ro) ro(SS-A) sjoegren syndrome type A antigen tripartite motif-containing 21 tripartite motif-containing protein 21; Ro(SS-A); Sicca syndrome antigen A; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SS-A; tripartite motif-containing 21; Tripartite motif-containing protein 21
Gene Aliases: RNF81; Ro/SSA; RO52; SSA; SSA1; TRIM21
UniProt ID: (Human) P19474
Entrez Gene ID: (Human) 6737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.