Invitrogen
Manufacturer Code:PA5114483
Catalog # PIPA5114483
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Functional Assay (FN) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Anti-TRIM21 antibody was raised against a peptide corresponding to 16 amino acids near the center of human TRIM21. The immunogen is located within amino acids 290-340 of TRIM21. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa ribonucleoprotein autoantigen Ro/SS-A 52 kDa Ro protein 52kD Ro/SSA autoantigen E3 ubiquitin-protein ligase TRIM21 RING finger protein 81 RING-type E3 ubiquitin transferase TRIM21 RNF81 Ro(SS-A) Ro/SSA RO52 Sicca syndrome antigen A sjoegren syndrome type A antigen Sjogren syndrome antigen A1 Sjogren syndrome antigen A1 (52kDa ribonucleoprotein autoantigen SS-A/Ro) SSA SS-A Ssa1 TRIM21 tripartite motif containing 21 tripartite motif protein 21 tripartite motif-containing 21 tripartite motif-containing protein 21; 52 kDa Ro protein; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; Ro(SS-A); Sicca syndrome antigen A; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SS-A; tripartite motif-containing 21; Tripartite motif-containing protein 21
Gene Aliases: RNF81; Ro/SSA; RO52; SSA; SSA1; TRIM21
UniProt ID: (Human) P19474
Entrez Gene ID: (Human) 6737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.