Invitrogen
Manufacturer Code:PA540946
Catalog # PIPA540946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TRIM15 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RING finger protein 93; Tripartite motif containing 15 RNF93 ZNFB7 ZNF178; tripartite motif protein 15; tripartite motif-containing 15; Tripartite motif-containing protein 15; Zinc finger protein 178; Zinc finger protein B7
Gene Aliases: 1810012B10Rik; RNF93; TRIM15; ZNF178; ZNFB7
UniProt ID: (Human) Q9C019
Entrez Gene ID: (Rat) 100910806, (Bovine) 540074, (Human) 89870, (Rabbit) 100348486, (Mouse) 69097
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.