Invitrogen
Manufacturer Code:MA526477
Catalog # PIMA526477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 120-420 of TICAM1 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB TICAM-1 TRIF TIR domain containing adaptor inducing interferon-beta TIR domain-containing adapter molecule 1 TIR domain-containing adapter protein inducing IFN-beta proline-rich vinculin and TIR domain-containing protein B putative NF-kappa-B-activating protein 502H toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; Putative NF-kappa-B-activating protein 502H; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; toll-interleukin I receptor domain containing adaptor molecule 1; Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1
Gene Aliases: IIAE6; MyD88-3; PRVTIRB; TICAM-1; TICAM1; TRIF
UniProt ID: (Human) Q8IUC6
Entrez Gene ID: (Human) 148022, (Rhesus monkey) 574294
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.