Invitrogen
Manufacturer Code:PA595104
Catalog # PIPA595104
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of mouse TRIF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TICAM-1; TIR domain-containing adapter molecule 1; TIR domain-containing adapter molecule 1 TICAM-1 Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta TIR domain-containing adapter protein inducing IFN-beta Ticam1 Trif; TIR domain-containing adapter protein inducing IFN-beta; TIR domain-containing adaptor inducing interferon-beta; toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta; Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta
Gene Aliases: AW046014; AW547018; TICAM-1; Ticam1; TRIF
UniProt ID: (Mouse) Q80UF7
Entrez Gene ID: (Mouse) 106759
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.