Invitrogen
Manufacturer Code:PA528105
Catalog # PIPA528105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 650 and 712 of Human TICAM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB TICAM-1 TRIF TIR domain containing adaptor inducing interferon-beta TIR domain-containing adapter molecule 1 TIR domain-containing adapter protein inducing IFN-beta proline-rich vinculin and TIR domain-containing protein B putative NF-kappa-B-activating protein 502H; Putative NF-kappa-B-activating protein 502H; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1
Gene Aliases: IIAE6; MyD88-3; PRVTIRB; TICAM-1; TICAM1; TRIF
UniProt ID: (Human) Q8IUC6
Entrez Gene ID: (Human) 148022
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.