Abnova
Manufacturer Code:PAB0336
Catalog # 89-114-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 315-332 of human TRIB3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TRIB3.
For Research Use Only
Protein Aliases: C20orf97; neuronal cell death inducible putative kinase; Neuronal cell death-inducible putative kinase; NIPK; p65-interacting inhibitor of NF-kappa-B; p65-interacting inhibitor of NF-kappaB; SINK; SKIP3; TRB-3; TRB3; Tribbles homolog 3
Gene Aliases: C20orf97; NIPK; SINK; SKIP3; TRB3; TRIB3
UniProt ID: (Human) Q96RU7
Entrez Gene ID: (Human) 57761
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.