Proteintech
Manufacturer Code:153591AP150UL
Catalog # 50-173-0176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIB2 Fusion Protein Ag7605 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C5FW GS3955 TRB 2 TRB2 TRIB2 Tribbles homolog 2; TRB-2; TRIB2; Tribbles homolog 2
Gene Aliases: AW319517; C5FW; GS3955; RGD1564451; TRB-2; TRB2; TRIB2
UniProt ID: (Human) Q92519, (Mouse) Q8K4K3
Entrez Gene ID: (Human) 28951, (Rat) 313974, (Mouse) 217410
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.