Proteintech
Manufacturer Code:153511AP150UL
Catalog # 50-173-0169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRIAP1 Fusion Protein Ag7597 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 15E1.1 HSPC132 P53CSV Protein 15E1.1 TRIAP1 WF 1; mitochondrial distribution and morphology 35 homolog; p53-inducible cell-survival factor; p53CSV; Protein 15E1.1; TP53-regulated inhibitor of apoptosis 1; WF-1
Gene Aliases: 15E1.1; 1810015M01Rik; AU020874; AU043831; HSPC132; MDM35; P53CSV; TRIAP1; WF-1
UniProt ID: (Human) O43715, (Mouse) Q9D8Z2
Entrez Gene ID: (Human) 51499, (Rat) 691278, (Mouse) 69076
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.