Invitrogen
Manufacturer Code:MA531925
Catalog # PIMA531925
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H6B4 |
Immunogen | Purified recombit fragment of human TERF2 (AA: 324-500) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Telomeric DNA-binding protein; telomeric repeat binding protein 2; Telomeric repeat-binding factor 2; TRF2 TRBF2; TTAGGG repeat-binding factor 2
Gene Aliases: TERF2; TRBF2; TRF2
UniProt ID: (Human) Q15554
Entrez Gene ID: (Human) 7014
Molecular Function:
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.