Novus Biologicals
Manufacturer Code:NBP141219
Catalog # NBP141219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8A5 |
Immunogen | The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: NIMA-interacting protein 2; NIMA-interacting protein 2 PIN2hTRF1-AS Telomeric protein Pin2/TRF1 telomeric repeat binding factor (NIMA-interacting) 1 telomeric repeat binding factor 1 telomeric repeat binding protein 1 telomeric repeat-binding factor 1 TERF1 TRBF1 TRF1FLJ41416 TRFt-TRF1 TTAGGG repeat-binding factor 1; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; Telomeric repeat-binding factor 1; TTAGGG repeat-binding factor 1
Gene Aliases: hTRF1-AS; PIN2; t-TRF1; TERF1; TRBF1; TRF; TRF1
UniProt ID: (Human) P54274
Entrez Gene ID: (Human) 7013
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.