R&D Systems
Manufacturer Code:AF1828SP
Catalog # AF1828SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TREM-2 His19-Ser174 Accession # Q9NZC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: PLOSL2 TREM-2 Trem2a Trem2b Trem2c TREM-2triggering receptor expressed on myeloid cells 2a Triggering receptor expressed on monocytes 2 triggering receptor expressed on myeloid cells 2; TREM-2; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a
Gene Aliases: TREM-2; TREM2; Trem2a; Trem2b; Trem2c
UniProt ID: (Human) Q9NZC2
Entrez Gene ID: (Human) 54209
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.