Invitrogen
Manufacturer Code:MA516765
Catalog # PIMA516765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Functional Assay (FN) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | L5-B8.2A12.3A12 |
Immunogen | Mouse TREM-1 Fc fusion protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD354; TREM-1; triggering receptor expressed in monocytes 1; Triggering receptor expressed on myeloid cells 1
Gene Aliases: Trem1
UniProt ID: (Mouse) Q9JKE2
Entrez Gene ID: (Mouse) 58217
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.