Novus Biologicals
Manufacturer Code:NBP276531
Catalog # NBP276531
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL7111 |
Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids: KLTEEKYELKEGQTLDVK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: CD354; CD354 antigen TREM-1Triggering receptor expressed on monocytes 1 triggering receptor expressed on myeloid cells 1 triggering-receptor TREM1; TREM-1; Triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells 1
Gene Aliases: CD354; TREM-1; TREM1
UniProt ID: (Human) Q9NP99
Entrez Gene ID: (Human) 54210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.