Novus Biologicals
Manufacturer Code:NB11041535G
Catalog # NB11041535G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an N-terminal region of human TREK 1 (within residues 1-100). [Swiss-Prot# Q9NRT2] |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K2P2.1 potassium channel; MGC126742 MGC126744 potassium channel subfamily K member 2 subfamily K member 2 tandem-pore-domain potassium channel TREK-1 TREK-1 K(+) channel subunit TWIK-related potassium channel 1 two-pore potassium channel 1; Outward rectifying potassium channel protein TREK-1; Potassium channel subfamily K member 2; potassium channel subfamily k member 2 variant 1; potassium channel subfamily k member 2 variant 2; potassium channel, two pore domain subfamily K, member 2; potassium inwardly-rectifying channel, subfamily K, member 2; tandem-pore-domain potassium channel TREK-1; TREK-1 K(+) channel subunit; TWIK-related potassium channel 1; Two pore domain potassium channel TREK-1; Two pore potassium channel TPKC1; two-pore potassium channel 1
Gene Aliases: hTREK-1c; hTREK-1e; K2p2.1; KCNK2; TPKC1; TREK; TREK-1; TREK1
UniProt ID: (Human) O95069
Entrez Gene ID: (Human) 3776
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.