Invitrogen
Manufacturer Code:PA558981
Catalog # PIPA558981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TREH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha,alpha-trehalase; Alpha,alpha-trehalose glucohydrolase; TRE TREA alphaalpha-trehalose glucohydrolase trehalase; Trehalase; trehalase (brush-border membrane glycoprotein)
Gene Aliases: TRE; TREA; TREH
UniProt ID: (Human) O43280
Entrez Gene ID: (Human) 11181
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.