Invitrogen
Manufacturer Code:PA545708
Catalog # PIPA545708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TREH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha,alpha-trehalase; Alpha,alpha-trehalose glucohydrolase; Trehalase; trehalase (brush-border membrane glycoprotein)
Gene Aliases: 2210412M19Rik; si:ch211-147p17.2; TRE; TREA; TREH
UniProt ID: (Rabbit) P19813, (Human) O43280, (Mouse) Q9JLT2
Entrez Gene ID: (Rabbit) 100009126, (Human) 11181, (Bovine) 522889, (Horse) 100071276, (Dog) 489380, (Mouse) 58866, (Guinea pig) 100726401, (Rat) 60576, (Zebrafish) 795901
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.