Invitrogen
Manufacturer Code:PA522076
Catalog # PIPA522076
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 7 and 285 of Trehalase (Uniprot IDNo.O43280) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha,alpha-trehalase; Alpha,alpha-trehalose glucohydrolase; TRE TREA alphaalpha-trehalose glucohydrolase trehalase; Trehalase; trehalase (brush-border membrane glycoprotein)
Gene Aliases: 2210412M19Rik; TRE; TREA; TREH
UniProt ID: (Human) O43280, (Mouse) Q9JLT2
Entrez Gene ID: (Human) 11181, (Mouse) 58866
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.