Abnova
Manufacturer Code:MAB4526
Catalog # 89-111-872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TRIM-04 |
Immunogen | Recombinant protein corresponding to partial human TRAT1. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store in the dark at 4°C. Do not freeze.<br />Avoid prolonged exposure to light.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant TRAT1.
For Research Use Only
Protein Aliases: HSPC062; pp29/30; T cell receptor interacting molecule; T-cell receptor-associated transmembrane adapter 1; T-cell receptor-interacting molecule; TCRIM; TRIM
Gene Aliases: HSPC062; pp29/30; TCRIM; TRAT1; TRIM
UniProt ID: (Human) Q6PIZ9
Entrez Gene ID: (Human) 50852
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.