Proteintech
Manufacturer Code:132791AP150UL
Catalog # 50-172-8620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRAT1 Fusion Protein Ag4023 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HSPC062 pp29/30 TCRIM TRAT1 TRIM; pp29/30; T cell receptor associated transmembrane adaptor 1; T cell receptor interacting molecule; T-cell receptor interacting molecule; T-cell receptor-associated transmembrane adapter 1; T-cell receptor-interacting molecule; TRIM
Gene Aliases: C030046M14Rik; HSPC062; pp29/30; TCRIM; TRAT1; TRIM
UniProt ID: (Human) Q6PIZ9, (Mouse) Q3UU67
Entrez Gene ID: (Human) 50852, (Mouse) 77647, (Rat) 498075
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.