OriGene
Manufacturer Code:CF505629
Catalog # 50-166-3791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B5 |
Immunogen | Full length human recombit protein of human TRAPPC4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CGI-104 HSPC172 PTD009 SBDN SYNBINDIN TRS23; Hematopoietic stem/progenitor cell protein 172; Synbindin; Trafficking protein particle complex subunit 4; TRS23 homolog
Gene Aliases: CGI-104; HSPC172; PTD009; SBDN; SYNBINDIN; TRAPPC4; TRS23
UniProt ID: (Human) Q9Y296
Entrez Gene ID: (Human) 51399
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.