Invitrogen
Manufacturer Code:MA521923
Catalog # PIMA521923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D5 |
Immunogen | TRAPPC4 (AAH10866 1 a.a. &bsim 219 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietic stem/progenitor cell protein 172; Synbindin; Synbindin TRS23 homolog Hematopoietic stem/progenitor cell protein 172 SBDN; Trafficking protein particle complex subunit 4; TRS23 homolog
Gene Aliases: CGI-104; HSPC172; PTD009; SBDN; SYNBINDIN; TRAPPC4; TRS23
UniProt ID: (Human) Q9Y296
Entrez Gene ID: (Human) 51399
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.