Proteintech
Manufacturer Code:120741AP150UL
Catalog # 50-172-7692
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRAPPC4 Fusion Protein Ag2737 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 104 HSPC172 PTD009 SBDN SYNBINDIN TRAPPC4 TRS23 TRS23 homolog; Hematopoietic stem/progenitor cell protein 172; Synbindin; syndecan binding protein 2; Trafficking protein particle complex subunit 4; TRS23 homolog
Gene Aliases: 1500017G03Rik; AI303617; CGI-104; HSPC172; PTD009; Sbd; SBDN; Sdcbp2; SYNBINDIN; TRAPPC4; TRS23
UniProt ID: (Human) Q9Y296, (Rat) Q69BT7, (Mouse) Q9ES56
Entrez Gene ID: (Human) 51399, (Rat) 367073, (Mouse) 60409
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.