Novus Biologicals
Manufacturer Code:NB1002574AF594
Catalog # N102574A594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1525 and the C-terminus (residue 1581) of human Cofactor Required for Sp1 Transcriptional Activation 1using the numbering given in entry NP_004765.2 (GeneID 5469). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: Activator-recruited cofactor 205 kDa component; Activator-recruited cofactor 205 kDa component ARC205 CRSP1TRIP2 CRSP200p53 regulatory protein RB18A DRIP230PPAR-binding protein mediator complex subunit 1DRIP205 mediator of RNA polymerase II transcription subunit 1 PBPPPARGBP Peroxisome proliferator-activated receptor-binding protein PPAR binding protein PPARBPMGC71488 PPARG binding protein RB18ATR-interacting protein 2 Thyroid hormone receptor-associated protein complex 220 kDa component thyroid hormone receptor-associated protein complex component TRAP220 thyroid receptor interacting protein 2 Thyroid receptor-interacting protein 2 Trap220 TRAP220TRIP-2 vitamin D receptor-interacting protein 230 kD Vitamin D receptor-interacting protein complex component DRIP205; ARC205; Mediator complex subunit 1; Mediator of RNA polymerase II transcription subunit 1; p53 regulatory protein RB18A; PBP; Peroxisome proliferator-activated receptor-binding protein; PPAR binding protein; PPAR-binding protein; PPARG binding protein; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; Thyroid receptor-interacting protein 2; TR-interacting protein 2; Trap220; TRIP-2; vitamin D receptor-interacting protein 230 kD; Vitamin D receptor-interacting protein complex component DRIP205
Gene Aliases: ARC205; CRSP1; CRSP200; DRIP205; DRIP230; MED1; PBP; PPARBP; PPARGBP; RB18A; TRAP220; TRIP2
UniProt ID: (Human) Q15648
Entrez Gene ID: (Human) 5469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.