R&D Systems
Manufacturer Code:AF5520
Catalog # AF5520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human TRAP220 Phe878-Phe1031 Accession # Q15648 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator-recruited cofactor 205 kDa component; Activator-recruited cofactor 205 kDa component ARC205 CRSP1TRIP2 CRSP200p53 regulatory protein RB18A DRIP230 DRIP230PPAR-binding protein MBD4 MED1 mediator complex subunit 1DRIP205 mediator of RNA polymerase II transcription subunit 1 PBPPPARGBP Peroxisome proliferator-activated receptor-binding protein PPAR binding protein PPARBP PPARBPMGC71488 PPARG binding protein PPARGBP RB18ATR-interacting protein 2 Thyroid hormone receptor-associated protein complex 220 kDa component thyroid hormone receptor-associated protein complex component TRAP220 thyroid receptor interacting protein 2 Thyroid receptor-interacting protein 2 Trap220 TRAP220TRIP-2 TRIP2 vitamin D receptor-interacting protein 230 kD Vitamin D receptor-interacting protein complex component DRIP205; ARC205; Mediator complex subunit 1; Mediator of RNA polymerase II transcription subunit 1; p53 regulatory protein RB18A; PBP; Peroxisome proliferator-activated receptor-binding protein; PPAR binding protein; PPAR-binding protein; PPARG binding protein; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; Thyroid receptor-interacting protein 2; TR-interacting protein 2; Trap220; TRIP-2; vitamin D receptor-interacting protein 230 kD; Vitamin D receptor-interacting protein complex component DRIP205
Gene Aliases: ARC205; CRSP1; CRSP200; DRIP205; DRIP230; MED1; PBP; PPARBP; PPARGBP; RB18A; TRAP220; TRIP2
UniProt ID: (Human) Q15648
Entrez Gene ID: (Human) 5469
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.