Invitrogen
Manufacturer Code:PA536114
Catalog # PIPA536114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 1420-1470 of human TRAP220 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activator-recruited cofactor 205 kDa component; ARC205; Mediator complex subunit 1; Mediator of RNA polymerase II transcription subunit 1; Mediator of RNA polymerase II transcription subunit 1 Activator-recruited cofactor 205 kDa component ARC205 Mediator complex subunit 1 Peroxisome proliferator-activated receptor-binding protein PBP PPAR-binding protein Thyroid hormone receptor-associated protein complex 220 kDa component Trap220 Thyroid receptor-interacting protein 2 TR-interacting protein 2 TRIP-2 Vitamin D receptor-interacting protein complex component DRIP205 p53 regulatory protein RB18A MED1 ARC205 CRSP1 CRSP200 DRIP205 DRIP230 PBP PPARBP PPARGBP RB18A TRAP220 TRIP2; p53 regulatory protein RB18A; PBP; peroxisome proliferator activated receptor binding protein; Peroxisome proliferator-activated receptor-binding protein; PPAR binding protein; PPAR-binding protein; PPARG binding protein; Thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; Thyroid receptor-interacting protein 2; TR-interacting protein 2; Trap220; TRIP-2; vitamin D receptor-interacting protein 230 kD; Vitamin D receptor-interacting protein complex component DRIP205
Gene Aliases: AI480703; ARC205; CRSP1; CRSP200; CRSP210; DRIP205; DRIP230; l11Jus15; MED1; PBP; PPARBP; PPARGBP; RB18A; TRAP220; TRIP2
UniProt ID: (Human) Q15648, (Mouse) Q925J9
Entrez Gene ID: (Human) 5469, (Mouse) 19014
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.