OriGene
Manufacturer Code:TA504202
Catalog # 50-167-4938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H8 |
Immunogen | Full length human recombit protein of human TRAP1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Heat shock protein 75 kDa, mitochondrial; HSP 75; HSP 75 HSP75 HSP90L TRAP-1; testicular tissue protein Li 209; TNF receptor-associated protein 1; TNFR-associated protein 1; TRAP-1; tumor necrosis factor type 1 receptor associated protein; Tumor necrosis factor type 1 receptor-associated protein
Gene Aliases: HSP 75; HSP75; HSP90L; TRAP-1; TRAP1
UniProt ID: (Human) Q12931, (Rat) Q5XHZ0
Entrez Gene ID: (Dog) 102153194, (Human) 10131, (Rat) 287069
Molecular Function:
Hsp90 family chaperone
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.